BAF420740115
SP Bel-Art® Secador® 4.0 Horizontal Auto-Desiccator Cabinet
internal volume 1.9 ft3, AC/DC input 120 V, US 3-pin plug, clear polyester (Co-polyester)
Synonym(s):
SECADOR 4.0 HORIZONTAL AUTO-DESICCATOR CABINETS, SP BEL-ART SECADOR CLEAR 4.0 HORIZONTAL AUTO-DESICCATOR CABINET; 120V, 1.9 CU. FT.
About This Item
Recommended Products
material
clear polyester (Co-polyester)
polycarbonate
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F42074-0115
internal volume
1.9 ft3
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Polycarbonate construction blocks 99% of UV light and resists staining, crazing and chemical attack
- Amber color models reduce visible light penetration by over 50%; perfect for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Color-indicating silica gel inside the desiccant regeneration module allows quick visual confirmation of desiccant status
- Large, pull down, oven style door with easy-to-view digital hygrometer maximizes access to interior space; includes patented seal design, tab latches and security loops for a padlock or tamper-evident seals; opens right to left
- Two removable, perforated shelves facilitate circulation of dry air
- Choose from a selection of sizes, styles and colors in 100, 120 or 230V (CE marked) models for use worldwide
- 230V units include power cord/transformer that includes Q-NA, Q-SAA, Q-UK and Q-EU input connectors to accommodate North American, Australian, UK and European configurations
- Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
- Use BAF420740020 replacement desiccant for Secador® Auto Desiccator Module
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service