Skip to Content
Merck
All Photos(1)

Key Documents

Z407259

Supelco

Aldrich® TLC developing tank - cylindrical

tank lid

Synonym(s):

Cylindrical TLC Tank

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$52.30

$52.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$52.30

About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

$52.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

technique(s)

thin layer chromatography (TLC): suitable

Looking for similar products? Visit Product Comparison Guide

General description

Can be used as a staining chamber. Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the product number for purchasing the tank lid separately for the Aldrich® TLC developing tank - cylindrical?

    1 answer
    1. The developing tanks come with the lids and are available in two separate sizes:
      1. Z243906 - Aldrich® TLC developing tank - cylindrical, glass tank, O.D. × H 6.5 cm × 10.5 cm
      2. Z243914 - Aldrich® TLC developing tank - cylindrical, glass tank, O.D. × H 6.5 cm × 21.0 cm
      To purchase the tank lid by itself, it would be product Z407259.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service