Skip to Content
Merck
All Photos(1)

Key Documents

Z204226

Supelco

Rectangular TLC developing tanks, complete

L × H × W 7.5 cm × 15.5 cm × 8.0 cm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$141.00

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$141.00

About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

technique(s)

thin layer chromatography (TLC): suitable

L × H × W

7.5 cm × 15.5 cm × 8.0 cm

Looking for similar products? Visit Product Comparison Guide

Footnote

TLC Developing Tanks come complete with the developing tank and lid

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service