Skip to Content
Merck
All Photos(1)

Key Documents

SYNT216105

Synthware TLC cylindrical developing tank

joint: ST/NS 60/12, O.D. × H 65 mm × 105 mm

Synonym(s):

Synthware T216105

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$171.00

$171.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$171.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.51

$171.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Joint

joint: ST/NS60/12

packaging

pkg of 1 ea

manufacturer/tradename

Synthware T216105

O.D. × H

65 mm × 105 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service