Skip to Content
Merck
All Photos(6)

Documents

1.52610

Supelco

TLC Explorer

greener alternative

Digital TLC analysis and documentation device

Synonym(s):

Digitization instrument for TLC plate analysis, TLC plate analyzer, TLC plate reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.21

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-30°C

General description

The TLC Explorer Documentation System is designed to digitally evaluate and record thin-layer chromatography (TLC), boosting the efficiency and accuracy of chromatography workflows. It provides precise TLC imaging for reliable densitometric measurement, facilitating quantitative analysis and consistent data interpretation.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Features and Benefits

  • Digitalized solution converts spot intensity into numerical data, allowing for quantitative and qualitative analysis of TLC, HPTLC, and PLC plates.
  • Digital imaging in less than 2 minutes allows for consistent capture of TLC plates, reducing subjectivity. The software analyzes spots based on pixel intensity and location, leading to more reliable interpretations.
  • Automation and digitalization help standardize the TLC process, reducing variations caused by human factors. This leads to greater consistency in analysis and improved reproducibility.
  • Enhanced documentation and archiving of digital records for saving, sharing, and retrieving. Researchers can track historical data and compare results over time more efficiently.

Other Notes

USER-FRIENDLY DESIGN AND OPERATION

The software enables TLC plate analysis in under 2 minutes, with automated track recognition and simultaneous measurement of multiple plates across a 20 x 20 cm area. It includes features like auto-image rotation for misaligned plates, background noise reduction filters, and automatic detection of the starting line and solvent front, with manual adjustment options. Annotations can be added for extra clarity, and the software supports both comparative and quantitative examinations of plates.

The device uses three LED light sources—white light (VIS), UV-A (366 nm), and UV-C (254 nm) — to illuminate the plates from above in direct-light mode and identify distinct substances. It features glove-compatible touch sensors for easy operation, a camera, and a PC unit with an antenna, all designed to meet industrial standards (URS). Connecting to the system is easy via Wi-Fi or QR code scan for smartphones, laptops, or desktop operation. This plug-and-play setup requires no technical support. The instrument can also use a power bank (sold separately) for added portability.

ERGONOMIC VISUAL PLATE ANALYSIS

The TLC Explorer features an inspection window for safe and quick examination of the plate. A manual shutter slider controls light entry, allowing the user to adjust visibility and minimize stray light. It provides three types of excitation sources, easily switched on with a tap on the touch-sensitive buttons, indicating their wavelengths. However, only one light source can be active at a time. 

SAFETY AND SUSTAINABILITY

The device features a safety setup that prevents UV exposure by ensuring the analytical process only begins when the plates are secured and closed in the Drawer Unit. A “Drawer Open” light indicates that the drawer has not been closed properly. The instrument also uses energy-efficient LED modules instead of traditional mercury lamps for its light source, providing a more sustainable and durable option.

EASY MAINTENANCE AND LAMP CHANGE

The exterior of the TLC device is easy to clean with commonly used solvents. Regular service maintenance is not required. Replacing the lamp unit can be done with minimal effort. Replacement parts and accessories can be ordered separately. If you need any further assistance, please contact your local Tech Service or visit our service webpage: SigmaAldrich.com/TLCservice
WHAT IS INCLUDED:

  • TLC Explorer device including illumination unit and software
  • Baseplate for TLC plates
  • Power plugs for worldwide connection
  • Instruction Manual & "How to Start" Quick Guide
  • QR code sticker for quick connection
  • Dust cover


FREQUENTLY ASKED QUESTIONS (FAQS)

Can the software perform quantification or semi-quantitate analyses:


Yes, it can. The software can calculate concentrations by comparing the spot on a track with corresponding spots on reference tracks that have known concentrations.

Is the software tool GMP-compliant:

No, the software is not fully GMP-compliant.

Can the instrument analyze plate formats other than 20 x 10 cm and those with backings other than glass:


Yes, the instrument accommodates a variety of plate sizes and thicknesses for added versatility. It supports aluminum- or plastic-backed TLC, HPTLC or PLC plates with different formats and shapes, up to 20 x 20 cm.

How many different plates can be analyzed in parallel:


You can load several plates or sheets on the base plate, which has an area of 20 x 20 cm at the top of the drawer. The software can analyze multiple plates in parallel, accurately distinguishing between each one during the measurements.

Can I also analyze the HPTLC plates:


Yes, the instrument (including software) allows a highly reliable analysis of HPTLC, TLC, and PLC plates.

Can I run a wavelength scan:


No, unfortunately, this feature is not available. You can carry out excitations at 254 nm, 366 nm, and VIS wavelengths.

Is there a warranty included:


Yes, we offer a one-year warranty. And in case you encounter any technical issues, please contact your local representative for further advice.

What is the expected lifetime of the lamp:


The lamp unit is equipped with three LEDs, which have a significantly longer lifetime and are more sustainable than mercury lamps. The LEDs can last 2-7 years, depending on the hours of usage.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Qualitative and quantitative TLC analyses. Automated software for hassle-free, quick, and precise results. Plug-and-play setup with intuitive software.

Discover an efficient and automated software for accurate and reliable qualitative and quantitative TLC analyses. Enjoy a hassle-free plug-and-play experience and easily connect through WiFi or LAN for seamless connectivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service