Key Documents
Z265268
Supelco
Adsorbent scrapers
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
manufacturer/tradename
Analtech 0500
technique(s)
thin layer chromatography (TLC): suitable
General description
Ideal flat 13 mm steel blade tapered to a sharp edge for rapid removal of adsorbent from TLC plates. Aluminum handle; replaceable blade.
related product
Product No.
Description
Pricing
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)