Skip to Content
Merck
All Photos(1)

Key Documents

Z214337

Ace-Thred Thermometer/Tubing Adapter

With 2mm bore PTFE stopcock plug and hose connection, joint: ST/NS 24/40, thread I.D. 7.0 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$179.00

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$179.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

label

E

Joint

joint: ST/NS24/40

manufacturer/tradename

Ace Glass 527215

thread I.D.

7.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

Threaded nylon bushing with FETFE® O-ring.

Legal Information

Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.

replacement part

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service