Key Documents
SYNT230015
Synthware graduated distillation receiver tube with hooks
capacity 15 mL, joint: ST/NS 14/20
Synonym(s):
Synthware T230015, distilling receiver tube
Select a Size
1 EA
$48.60
$48.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$48.60
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$48.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
V-bottom bottom
colorless tube
glass tube
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware T230015
capacity
15 mL
General description
Centrifuge tube shaped receiver with an upper standard taper joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)