Key Documents
Z125350
Ace-Thred Thermometer/Tubing Adapter
Straight, joint: ST/NS 10/18, thread I.D. 6.5 mm
Select a Size
1 EA
$97.20
$97.20
Available to ship onApril 01, 2025Details
Select a Size
Change View
1 EA
$97.20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$97.20
Available to ship onApril 01, 2025Details
label
A
Joint
joint: ST/NS10/18
manufacturer/tradename
Ace Glass 502824
thread I.D.
6.5 mm
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)