Key Documents
Z274240
Sigma-Aldrich
Aldrich® tube fitting kit
for tubing i.d., 1/8 in.
Select a Size
1 KIT
$341.00
$341.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 KIT
$341.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$341.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
for tubing i.d. 1/8 in.
General description
Clear, natural Nylon-66 fittings for making a variety of connections with flexible tubing. Fittings are supplied in a compartmentalized PP storage box.
Contains 165 fittings that include: tubing hose barbs, connectors, tees, branched tees, elbows, plugs, reducers, and locking sleeves.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)