Skip to Content
Merck
All Photos(1)

Key Documents

W415655

Sigma-Aldrich

Menthyl valerate

natural (US), ≥98%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W415655-SAMPLE-K
$58.00
100 G
$290.00
500 G
$943.00

$58.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W415655-SAMPLE-K
$58.00
100 G
$290.00
500 G
$943.00

About This Item

Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
FEMA Number:
4156
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.154
grade:
FG
Halal
Kosher
natural (US)
food allergen:
no known allergens

$58.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Halal
Kosher
natural (US)

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; minty; sweet

SMILES string

Fc1c(c(c(c(c1F)F)C(=O)Nc2cc3c(cc2)C4(OC3O)c5c(cc(cc5)O)Oc6c4ccc(c6)O)F)F

InChI

1S/C27H14F5NO6/c28-20-19(21(29)23(31)24(32)22(20)30)25(36)33-10-1-4-14-13(7-10)26(37)39-27(14)15-5-2-11(34)8-17(15)38-18-9-12(35)3-6-16(18)27/h1-9,26,34-35,37H,(H,33,36)

InChI key

AJIMMVOZOJXJKD-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service