Skip to Content
Merck
All Photos(1)

Key Documents

W461216

Sigma-Aldrich

2-Ethyl-2-hexenal

mixture of isomers, natural (US), ≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

W461216-SAMPLE-K
$62.20
100 G
$139.00
1 KG
$1,050.00

$62.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W461216-SAMPLE-K
$62.20
100 G
$139.00
1 KG
$1,050.00

About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
FEMA Number:
4612
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Halal
Kosher
natural (US)
biological source:
botanical
food allergen:
no known allergens

$62.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

botanical

Quality Level

grade

Halal
Kosher
natural (US)

reg. compliance

FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; spicy; woody; minty

SMILES string

O=C\C(=C\CCC)\CC

InChI

1S/C8H14O/c1-3-5-6-8(4-2)7-9/h6-7H,3-5H2,1-2H3/b8-6+

InChI key

PYLMCYQHBRSDND-SOFGYWHQSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

309.9 °F

Flash Point(C)

154.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service