Skip to Content
Merck
All Photos(1)

Key Documents

W346500

Sigma-Aldrich

cis-6-Nonen-1-ol

≥95%, FG

Synonym(s):

(Z)-6-Nonen-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

W346500-SAMPLE-K
$54.60
100 G
$349.00
1 KG
$2,350.00

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W346500-SAMPLE-K
$54.60
100 G
$349.00
1 KG
$2,350.00

About This Item

Linear Formula:
C2H5CH=CH(CH2)5OH
CAS Number:
Molecular Weight:
142.24
FEMA Number:
3465
EC Number:
Council of Europe no.:
10294
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.093
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥95%

refractive index

n20/D 1.449 (lit.)

bp

115 °C/20 mmHg (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; melon; waxy

SMILES string

[H]\C(CC)=C(/[H])CCCCCO

InChI

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h3-4,10H,2,5-9H2,1H3/b4-3-

InChI key

XJHRZBIBSSVCEL-ARJAWSKDSA-N

Related Categories

General description

cis-6-Nonen-1-ol occurs naturally in melon fruits[1][2] such as honeydew[3], muskmelon[4] and watermelon.[5]

Application


  • RIFM fragrance ingredient safety assessment, cis-6-nonen-1-ol, CAS Registry Number 35854-86-5.: This safety assessment evaluates the use of cis-6-nonen-1-ol in fragrances, ensuring its safety for consumer use. The study underscores the compound′s potential applications in the fragrance industry, emphasizing its non-toxic nature and suitability for various consumer products (Api et al., 2017).


Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristics of fresh-cut honeydew (Cucumis xmelo L.) available to processors in winter and summer and its quality maintenance by modified atmosphere packaging
Bai J, et al.
Postharvest biology and technology, 97(1), 349-359 (2003)
Fast quantitative determination of aroma volatile constituents in melon fruits by headspace?solid-phase microextraction and gas chromatography?mass spectrometry
Verzera A, et al.
Food Analytical Methods, 28(3), 141-149 (2011)
Quality characteristics of fresh-cut watermelon slices from non-treated and 1-methylcyclopropene-and/or ethylene-treated whole fruit.
Saftner R, et al.
Postharvest biology and technology, 29(1), 65-67 (1981)
Comparison of the volatile compositions and flavour properties of cantaloupe, Galia and honeydew muskmelons
Kourkoutas D, et al.
Food Chemistry, 44(1), 71-79 (2007)
Comparison of characteristic flavor and aroma volatiles in melons and standards using solid phase microextraction (SPME) and stir bar sorptive extraction (SBSE) with GC-MS
Beaulieu JC, et al.
Hortscience: a Publication of the American Society For Horticultural Science Hortscience, 4(2), 2707-2707 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service