W481015
Ethyl-2-(4-hydroxy-3-methoxyphenyl)acetate
≥95%
Synonym(s):
Ethyl homovanillate, Ethyl 4-hydroxy-3-methoxyphenylacetate
Select a Size
$2,250.00
Select a Size
About This Item
$2,250.00
Recommended Products
biological source
synthetic
Quality Level
grade
Halal
reg. compliance
FDA 21 CFR 117
Assay
≥95%
bp
180-185 °C/14 mmHg (lit.)
mp
44-47 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
musty; spicy
SMILES string
CCOC(=O)Cc1ccc(O)c(OC)c1
InChI
1S/C11H14O4/c1-3-15-11(13)7-8-4-5-9(12)10(6-8)14-2/h4-6,12H,3,7H2,1-2H3
InChI key
AWPMWZHWVKXADV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
EPA's Twelve Principles of Green Chemistry guide Flavors and Fragrances industry toward environmentally friendly practices.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service