Skip to Content
Merck
All Photos(1)

Key Documents

T49409

Sigma-Aldrich

2,4,6-Tri-tert-butylphenol

98%

Synonym(s):

2,4,6-Tri-t-butylphenol, 2,4,6-Tri-tert-butyl-1-hydroxybenzene, 2,4,6-Tris(1,1-dimethylethyl)phenol, 2,4,6-Tris(tert-butyl)phenol, 2,4,6-Tritert-butylphenol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$163.00

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$163.00

About This Item

Linear Formula:
[(CH3)3C]3C6H2OH
CAS Number:
Molecular Weight:
262.43
Beilstein:
1913256
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

bp

277 °C (lit.)

mp

125-130 °C (lit.)

SMILES string

CC(C)(C)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C18H30O/c1-16(2,3)12-10-13(17(4,5)6)15(19)14(11-12)18(7,8)9/h10-11,19H,1-9H3

InChI key

PFEFOYRSMXVNEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Sens. 1B - STOT RE 2

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshinori Kadoma et al.
Chemosphere, 74(5), 626-632 (2008-12-17)
To clarify the mechanism of phenol toxicity, the radical-scavenging activity of 2- or 2,6-di-tert-butyl- and 2-methoxy-substituted phenols was investigated by combining two distinct approaches: first, the induction period method for methacrylate polymerization initiated by benzoyl peroxide or 2,2'-azobisisobutyronitrile; and secondly
A L Wilcox et al.
Chemical research in toxicology, 6(4), 413-416 (1993-07-01)
13-Hydroperoxyoctadeca-9,11,15-trienoic acid was reacted with a catalytic amount of 5,10,15,20-tetraphenyl-21H,23H-porphyrin iron(III) chloride in dichloromethane containing 2,4,6-tri-tert-butylphenol. The principal products were identified as 13-oxooctadeca-9,11,15-trienoic acid, 13-oxotrideca-9,11-dienoic acid, and a series of isomeric epoxyaryl ethers [9-(2,4,6-tri-tert-butylphenoxy)-12,13-epoxyoctadec-10-enoic acids and 11-(2,4,6-tri-tert-butylphenoxy)-12,13-epoxyoctadec-9-enoic acids]. The epoxyaryl
O Takahashi et al.
Xenobiotica; the fate of foreign compounds in biological systems, 13(5), 319-326 (1983-05-01)
Single oral doses of the haemorrhagic antioxidant 2,4,6-tri-t-butylphenol (260 mg/kg) were well absorbed in the rat. Peak blood levels of this compound were reached in 15-60 min. The blood elimination half-lives were 18.2 min for the alpha-phase and 11.8 h
S Nemoto et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 42(6), 359-366 (2002-03-06)
An analytical method has been developed for the determination of 2,4,6-tri-tert-butylphenol (TTBP) in foods. TTBP was determined by GC/MS (SIM) after extraction from food samples using a steam distillation technique. The developed method was able to determine simultaneously 2,4-di-tert-butylphenol (2,4-DTBP)
K Matsumoto et al.
The Journal of toxicological sciences, 16(4), 167-179 (1991-11-01)
In order to study the chronic toxicity of 2, 4, 6-tri-tert-butylphenol (TTBP), groups of 40 Slc: Wistar rats of either sex were fed diet containing 0, 30, 100, 300 or 1000 ppm of TTBP for up to 24 months. Hematological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service