Skip to Content
Merck
All Photos(2)

Key Documents

322806

Sigma-Aldrich

2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol

98%

Synonym(s):

Octrizole

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$326.00

$326.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$326.00

About This Item

Empirical Formula (Hill Notation):
C20H25N3O
CAS Number:
Molecular Weight:
323.43
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$326.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

98%

form

powder

mp

106-108 °C (lit.)

SMILES string

CC(C)(C)CC(C)(C)c1ccc(O)c(c1)-n2nc3ccccc3n2

InChI

1S/C20H25N3O/c1-19(2,3)13-20(4,5)14-10-11-18(24)17(12-14)23-21-15-8-6-7-9-16(15)22-23/h6-12,24H,13H2,1-5H3

InChI key

IYAZLDLPUNDVAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

UV absorber

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service