Skip to Content
Merck
All Photos(1)

Key Documents

B97607

Sigma-Aldrich

2-tert-Butyl-6-methyl-phenol

99%

Synonym(s):

6-tert-Butyl-o-cresol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$88.00

$88.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$88.00

About This Item

Linear Formula:
(CH3)3CC6H3(CH3)OH
CAS Number:
Molecular Weight:
164.24
Beilstein:
1862362
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$88.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

99%

form

crystals

refractive index

n20/D 1.519 (lit.)

bp

230 °C (lit.)

mp

24-27 °C (lit.)

density

0.967 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1O)C(C)(C)C

InChI

1S/C11H16O/c1-8-6-5-7-9(10(8)12)11(2,3)4/h5-7,12H,1-4H3

InChI key

BKZXZGWHTRCFPX-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerry O Ebalunode et al.
Bioorganic & medicinal chemistry, 17(14), 5133-5138 (2009-06-13)
Current anesthetics, especially the inhaled ones, have troublesome side effects and may be associated with durable changes in cognition. It is therefore highly desirable to develop novel chemical entities that reduce these effects while preserving or enhancing anesthetic potency. In
Cynthia D Selassie et al.
Journal of medicinal chemistry, 48(23), 7234-7242 (2005-11-11)
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service