Skip to Content
Merck
All Photos(4)

Key Documents

B99901

Sigma-Aldrich

4-tert-Butylphenol

99%

Synonym(s):

4-(1,1-Dimethylethyl)phenol, Butylphen, PTBP, p-tert-Butylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1817334
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
99%
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 70 °C)

Quality Level

Assay

99%

form

powder

bp

236-238 °C (lit.)

mp

96-101 °C (lit.)

density

0.908 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccc(O)cc1

InChI

1S/C10H14O/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11H,1-3H3

InChI key

QHPQWRBYOIRBIT-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Butylphenol is a phenol derivative. Its contact with skin may lead to leukoderma. It is widely used in the polymer industry.[1] Reaction of 4-tert-Butylphenol with mushroom tyrosinase has been reported to afford 4-t-butyl-o-benzoquinone and kinetics of this enzymatic reaction has been investigated.[2]

Application

4-tert-Butylphenol is suitable reagent used in kinetic study of hydroxylation of 4-tert-butylphenol by mushroom tyrosinase.[2] It may be used in the synthesis of calix[7]arene.[3]
4-tert-Butylphenol may be employed as carbon and energy supplement in the culture medium of Sphingobium fuliginis strains.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Thörneby-Andersson et al.
Pigment cell research, 13(1), 33-38 (2000-04-13)
Exposure of the skin to certain phenols or catechols such as 4-tert-butylphenol (TBP) and 4-tert-butylcatechol (TBC) may cause leukoderma. These substances are used in the polymer industry and numerous cases have been reported. Several theories of the mechanism for chemical
Calix [7] arene from 4-tert-butylphenol and formaldehyde.
Nakamoto Y and Ishida S-I.
Makromol. Chem., Rapid Commun., 3(10), 705-707 (1982)
J R Ros et al.
European journal of biochemistry, 222(2), 449-452 (1994-06-01)
The reaction between 4-tert-butylphenol (BuPhOH) and mushroom tyrosinase was investigated by following 4-tert-butyl-ortho-benzoquinone, whose high stability permits the reaction to be used as a model for the study of the monophenolase activity of tyrosinase. The system evolves to a pseudo-steady
Sonnich Meier et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 145(3), 420-430 (2007-03-09)
Offshore oil production releases large amounts of lipophilic compounds in produced water into the ocean. In 2004, 143 million m(3) produced water, containing approximately 13 tons of long-chain (>C(4)) alkylphenols (AP), was discharged from installations in the Norwegian sector of
J Pablo Lamas et al.
Analytical and bioanalytical chemistry, 394(8), 2231-2239 (2009-07-07)
A method for the determination of dimethyl fumarate (DMF), benzothiazole (BT) and tert-butylphenol (TBP) in desiccant and antimould agents employed for protecting consumer products from humidity and mould has been developed. The method is based on ultrasound-assisted extraction (UAE) followed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service