Key Documents
SYNA611922
Synthware straight vacuum adapter with glass stopcock
male joint: ST/NS 19/22
Synonym(s):
Synthware A611922, glass adapter with stopcock
Select a Size
1 EA
$125.00
$125.00
Estimated to ship onMay 28, 2025
Select a Size
Change View
1 EA
$125.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$125.00
Estimated to ship onMay 28, 2025
Joint
male joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A611922
General description
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)