Key Documents
SYNA611420
Synthware straight vacuum adapter with glass stopcock
male joint: ST/NS 14/20
Synonym(s):
Synthware A611420, glass adapter with stopcock
Select a Size
1 EA
$121.00
$121.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$121.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$121.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
male joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A611420
Related Categories
General description
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)