Key Documents
SYNA702440
Synthware straight vacuum adapter with PTFE stopcock
male joint: ST/NS 24/40
Synonym(s):
Synthware A702440, flow control adapter
Select a Size
1 EA
$167.00
$167.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$167.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$167.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
male joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A702440
Related Categories
General description
Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. With standard taper inner joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)