Key Documents
SYNA701922
Synthware straight vacuum adapter with PTFE stopcock
male joint: ST/NS 19/22
Synonym(s):
Synthware A701922, flow control adapter
Select a Size
1 EA
$156.00
$156.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$156.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$156.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
male joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A701922
Related Categories
General description
Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. With standard taper inner joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)