Key Documents
HAM238160
Hamilton® Single Pore lab pH electrode
glass, S7 connector
Select a Size
1 EA
$491.00
$491.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$491.00
About This Item
UNSPSC Code:
41115604
NACRES:
NA.23
$491.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
cylindrical H glass membrane
glass
single pore diaphragm
feature
autoclavable: no
electrode Skylyte-CL electrolyte
S7 connector
parameter
0-100 °C temp. range
diam.
12 mm
pH
0-14
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)