Skip to Content
Merck
All Photos(1)

Key Documents

Z264946

Sigma-Aldrich

Aldrich® glass pH electrode

BNC connector

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$218.00

$218.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$218.00

About This Item

UNSPSC Code:
41115603
NACRES:
NB.79

$218.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

parameter

−5-110 °C temperature

Looking for similar products? Visit Product Comparison Guide

General description

For applications where a separate reference electrode is required. Fast response over the full pH range with low sodium ion error. Useful for highly acidic or highly alkaline samples.

Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Is the electrode "Z264946" refillable?

    1 answer
    1. Yes, it can be re-filled.

      Helpful?

  2. Please provide user guide for Z264946 Aldrich® glass pH electrode.

    1 answer
    1. A user guide is not available for this product. See the usage instructions below:
      PREPARATION FOR USE:
      1. Wet lower end of electrode in tap water. Carefully remove the lower cap.
      NOTE: The cap is used only for shipping and storage. You might want to keep it for later use.
      2. If air bubbles are present in the bulb area, shake downwards to displace any air bubbles in the bulb.
      pH MEASUREMENT:
      1. Thoroughly rinse the electrode tip in distilled water and insert electrode tip in a known buffer, close to the sample to be tested, along with an appropriate reference electrode.
      2. Adjust the ‘standardize’ control on the pH meter until the meter reads the value of the buffer.
      3. Thoroughly rinse the electrodes with distilled water, then insert the electrodes in the solution to be tested.
      NOTE: The electrode tips should be rinsed in distilled water between each measurement to prevent contamination of the next solution.
      STORAGE PROCEDURES:
      1. For short storage (up to one week), the electrode may be placed in a beaker containing approximately 1 inch saturated KCl solution.
      2. For long-term storage (over one week), fill storage cap with KCl solution and place the cap back on the electrode tip.
      The electrode may then be placed back in the box and stored.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service