Key Documents
Z113077
Sigma-Aldrich
Aldrich® glass pH electrode
Standard pin connector
Select a Size
1 EA
$253.00
$253.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$253.00
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
$253.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
parameter
−5-110 °C temperature
General description
For applications where a separate reference electrode is required. Fast response over the full pH range with low sodium ion error. Useful for highly acidic or highly alkaline samples.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)