Skip to Content
Merck
All Photos(1)

Key Documents

D173401

Sigma-Aldrich

2,4-Dimethylpentane

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$158.00
25 G
$446.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$158.00
25 G
$446.00

About This Item

Linear Formula:
(CH3)2CHCH2CH(CH3)2
CAS Number:
Molecular Weight:
100.20
Beilstein:
1696855
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

3.48 (vs air)

Quality Level

vapor pressure

3.29 psi ( 37.7 °C)

Assay

99%

form

liquid

expl. lim.

~7 %

refractive index

n20/D 1.381 (lit.)

bp

80 °C (lit.)

mp

−123 °C (lit.)

density

0.673 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)C

InChI

1S/C7H16/c1-6(2)5-7(3)4/h6-7H,5H2,1-4H3

InChI key

BZHMBWZPUJHVEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaiho Cho et al.
Chemosphere, 68(2), 266-273 (2007-02-16)
The biodegradability of alkylate compounds in serum bottles was investigated in the presence and absence of ethanol or benzene, toluene, ethylbenzene, and p-xylene (BTEX). The biomass was acclimated to three different alkylates, 2,3-dimethylpentane, 2,4-dimethylpentane and 2,2,4-trimethylpentane in porous pot reactors.
M B Snipes et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 16(1), 81-91 (1991-01-01)
This paper describes apparatus and methods for measuring uptake of inhaled vapors in the nose and lungs of dogs. The system allows sampling of air from the trachea at specific times during inspiration and expiration without surgical manipulation of the
A R Dahl et al.
Toxicology and applied pharmacology, 109(2), 263-275 (1991-06-15)
The site of uptake of inhaled vapors profoundly influences which respiratory tract tissues receive the highest doses. How the site of uptake depends on the physicochemical properties of inhaled vapors has been the subject of experiment and speculation for decades

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service