Skip to Content
Merck
All Photos(1)

Key Documents

M49801

Sigma-Aldrich

3-Methylhexane

99%

Synonym(s):

(±)-3-Methylhexane, 2-Ethylpentane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$150.00
25 G
$558.00

$150.00


Estimated to ship onApril 02, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$150.00
25 G
$558.00

About This Item

Linear Formula:
CH3CH2CH2CH(CH3)CH2CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1718739
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$150.00


Estimated to ship onApril 02, 2025


Request a Bulk Order

vapor pressure

2.13 psi ( 37.7 °C)

Quality Level

Assay

99%

form

liquid

autoignition temp.

536 °F

expl. lim.

~7 %

refractive index

n20/D 1.388 (lit.)

bp

91 °C (lit.)

mp

−119 °C (lit.)

density

0.687 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)CC

InChI

1S/C7H16/c1-4-6-7(3)5-2/h7H,4-6H2,1-3H3

InChI key

VLJXXKKOSFGPHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methylhexane can be used as a model alkane to study:
  • Cu-catalyzed hydrocarboxylation of simple alkanes to corresponding branched monocarboxylic acids in aqueous medium at low-temperature.[1]
  • Au-catalyzed oxygenation of alkanes with H2O2 in acetonitrile.[2]
  • FeCI3 catalyzed photooxygenation of simple alkanes to corresponding alcohols in presence of atmospheric O2.[3]

3-Methylhexane is one of the isomer of heptane.[4] It can be used as a reactant in the synthesis of dimethyl isomers (dimethyl pentanes) using Pt/WZA catalyst.[5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron (III) chloride catalysed photooxygenation of alcohol solutions of alkanes by atmospheric oxygen
Shul?pin GB and Druzhinina AN
Mendeleev Communications, 2(1), 36-37 (1992)
Mild, single-pot hydrocarboxylation of linear C5-C9 alkanes into branched monocarboxylic C6-C10 acids in copper-catalyzed aqueous systems
Kirillova MV, et al.
Applied Catalysis A: General, 401(1-2), 106-113 (2011)
Alumina-doped Pt/WOx/ZrO2 catalysts for n-heptane isomerization
Hua W and Sommer J
Applied Catalysis A: General, 232(1-2), 129-135 (2002)
Alkane oxygenation catalysed by gold complexes
Shul'pin GB, et al.
Tetrahedron Letters, 42(41), 7253-7256 (2001)
The influence of fuel structure on combustion as demonstrated by the isomers of heptane: a rapid compression machine study.
Silke EJ, et al.
Proceedings of the Combustion Institute, 30(2), 2639-2647 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service