Skip to Content
Merck
All Photos(1)

Key Documents

41085

Supelco

2,3-Dimethylpentane

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$263.00

$263.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$263.00

About This Item

Linear Formula:
C2H5CH(CH3)CH(CH3)2
CAS Number:
Molecular Weight:
100.20
Beilstein:
1718734
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$263.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3.45 (vs air)

vapor pressure

2.35 psi ( 37.7 °C)

Assay

≥99.0% (GC)

autoignition temp.

639 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

~7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.392 (lit.)
n20/D 1.392

bp

89-90 °C (lit.)

density

0.695 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCC(C)C(C)C

InChI

1S/C7H16/c1-5-7(4)6(2)3/h6-7H,5H2,1-4H3

InChI key

WGECXQBGLLYSFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dimethylpentane belongs to the class of volatile organic compounds (VOCs).[1]

Application

2,3-Dimethylpentane may be used as an analytical reference standard for the determination of 2,3-dimethylpentane in:
  • Ambient air samples by thermal desorption-gas chromatography (TD-GC) in conjunction with Deans-switch cutting technique.[2]
  • Exhaled air samples by GC combined with mass spectrometry (MS) operating under the selected ion monitoring (SIM) detection mode.[3]
  • Indoor and outdoor air samples by adsorption using sorbent tubes, followed by analysis using TD-GC-MS with electron impact (EI) ionization.[4]
  • Commercial reagent samples by high resolution GC using graphene-zeolite imdidazole framework (ZIF8) as a stationary phase.[5]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Influences of relative humidities and temperatures on the collection of C2-C5 aliphatic hydrocarbons with multi-bed (Tenax TA, Carbograph 1TD, Carboxen 1003) sorbent tube method
Ho SSH, et al.
Atmospheric Environment, 151, 45-51 (2017)
The application of chromatographic breath analysis in the search of volatile biomarkers of chronic kidney disease and coexisting type 2 diabetes mellitus.
Grabowska-Polanowska B, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1060, 103-110 (2017)
Graphene-ZIF8 composite material as stationary phase for high-resolution gas chromatographic separations of aliphatic and aromatic isomers
Yang X, et al.
Journal of Chromatography A, 1460, 173-180 (2016)
Optimization and evaluation of multi-bed adsorbent tube method in collection of volatile organic compounds.
Ho SSH, et al.
Atmospheric Research, 202, 187-195 (2018)
Characterization of thermal desorption with the Deans-switch technique in gas chromatographic analysis of volatile organic compounds.
Ou-Yang CF, et al.
Journal of Chromatography A, 1462, 107-114 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service