Skip to Content
Merck
All Photos(3)

Key Documents

300306

Sigma-Aldrich

Methylcyclohexane

anhydrous, ≥99%

Synonym(s):

Hexahydrotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$305.00
1 L
$619.00
2 L
$919.00

$305.00


Estimated to ship onApril 01, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$305.00
1 L
$619.00
2 L
$919.00

About This Item

Linear Formula:
C6H11CH3
CAS Number:
Molecular Weight:
98.19
Beilstein:
505972
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
101 °C (lit.)
vapor pressure:
37 mmHg ( 20 °C)
83.2 mmHg ( 37.7 °C)

$305.00


Estimated to ship onApril 01, 2025


Request a Bulk Order

grade

anhydrous

Quality Level

vapor density

3.4 (vs air)

vapor pressure

37 mmHg ( 20 °C)
83.2 mmHg ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

545 °F

expl. lim.

6.7 %

impurities

<0.002% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.422 (lit.)

bp

101 °C (lit.)

mp

−126 °C (lit.)

density

0.77 g/mL at 25 °C (lit.)

SMILES string

CC1CCCCC1

InChI

1S/C7H14/c1-7-5-3-2-4-6-7/h7H,2-6H2,1H3

InChI key

UAEPNZWRGJTJPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methylcyclohexane may be used as a solvent for the synthesis of unsolvated n-butylmagnesium chloride by the reaction of 1-chlorobutane with magnesium powder.[1] It may also be used as the starting material to synthesize 1-methylcyclohexanecarboxylic acid.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Won Baek et al.
Polymers, 11(7) (2019-06-30)
Hafnium complexes have drawn attention for their application as post-metallocene catalysts with unique performance in olefin polymerization. In this work, a series of half-metallocene HfMe2 complexes, bearing a tetrahydroquinoline framework, as well as a series of [Namido,N,Caryl]HfMe2-type post-metallocene complexes, bearing
Unsolvated n-butylmagnesium chloride
Bryce-Smith D and Blues ET
Organic Syntheses, 47, 113-113 (1967)
Kyung Lee Park et al.
Polymers, 12(5) (2020-05-15)
The pyridylamido hafnium complex (I) discovered at Dow is a flagship catalyst among postmetallocenes, which are used in the polyolefin industry for PO-chain growth from a chain transfer agent, dialkylzinc. In the present work, with the aim to block a
Thomas D Turner et al.
Journal of pharmaceutical sciences, 108(5), 1779-1787 (2018-12-28)
An analysis of the important intermolecular interactions of the active pharmaceutical ingredient lovastatin which contribute to the surface chemistry and attachment energy morphology is presented. The analysis is supported by a recent redetermination of the single-crystal structure (orthorhombic space group
1-Methylcyclohexanecarboxylic acid
Haaf W
Organic Syntheses, 46, 72-72 (1966)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service