CLS982516X
Pyrex® culture tubes with screw-cap
O.D. × L 16 mm × 125 mm
Synonym(s):
Corning® 9825
Select a Size
$332.00
Estimated to ship onMarch 24, 2025
Select a Size
About This Item
$332.00
Estimated to ship onMarch 24, 2025
Recommended Products
material
borosilicate glass
cap (Rubber Liner)
round bottom tube
thread size
15-415
sterility
non-sterile
packaging
pack of 144
case of 288
manufacturer/tradename
Corning 9825-16X
O.D. × L
16 mm × 125 mm
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service