CLS982516XX
Pyrex® culture tubes with screw-cap
O.D. × L 16 mm × 150 mm
Synonym(s):
Corning® 9825
Select a Size
$381.00
Select a Size
About This Item
$381.00
Recommended Products
material
borosilicate glass
cap (Rubber Liner)
round bottom tube
thread size
15-415
sterility
non-sterile
packaging
case of 288
pack of 96
manufacturer/tradename
Corning 9825-16XX
O.D. × L
16 mm × 150 mm
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service