Key Documents
CLS9999915
Corning® disposable phenolic cap
15-415 thread
Synonym(s):
phenolic caps, threaded cap, threaded caps
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
Pricing and availability is not currently available.
manufacturer/tradename
Corning 99999-15
H
16.0 mm
fitting
15-415 thread
General description
These disposable Corning caps are made from phenolic resin and are autoclavable at 121°C. Caps are provided with a white rubber liner.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)