CLS982525X
Pyrex® culture tubes with screw-cap
O.D. × L 25 mm × 200 mm
Synonym(s):
culture tube, glass tubes, test tubes, Corning® 9825
Select a Size
$751.00
Select a Size
About This Item
$751.00
Recommended Products
material
borosilicate glass
cap (Rubber Liner)
round bottom tube
thread size
24-410
sterility
non-sterile
packaging
case of 144
pack of 48
manufacturer/tradename
Corning 9825-25X
O.D. × L
25 mm × 200 mm
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service