Documents
BR65738
BRAND® desiccator ′Novus′ lid with socket NS 24/29
nominal diameter × O.D. × H 200 mm × 270 mm × 93 mm, joint: 24/29
Synonym(s):
Accessories, Lids
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Joint
joint: 24/29
feature
lid with socket
manufacturer/tradename
BRAND 65738
nominal diameter × O.D. × H
200 mm × 270 mm × 93 mm
General description
Crafted with high-quality materials, this lid features a robust knob for easy handling and secure sealing. Specifically designed for use with BRAND® glass desiccators, providing a perfect fit. Specifically designed for use with BRAND® glass desiccators, providing a perfect fit.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG