Skip to Content
Merck
All Photos(1)

Key Documents

Z740643

Sigma-Aldrich

Aldrich® Essentials desiccator

I.D. × H 150 mm × 250 mm, with perforated porcelain plate

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$424.00

$424.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$424.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

$424.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

I.D. × H

150 mm × 250 mm

Looking for similar products? Visit Product Comparison Guide

General description

Heavy-wall, molded, neutral glass, desiccators with ground flange top and lid. Lid has knob top to assist in removing lid. Complete with perforated, bottom, porcelain plate. No direct heat or flame can be used.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service