Key Documents
BR458732
BRAND® filter crucible
borosilicate glass, capacity 50 mL, 2 D 2
Synonym(s):
Filter, Filter crucible
Select a Size
10 EA
$586.00
$586.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
Select a Size
Change View
10 EA
$586.00
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
$586.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
material
borosilicate glass
manufacturer/tradename
BRAND 458732
capacity
50 mL
pore size
40-100 μm
General description
The BRAND® Filter Crucible made from Boro 3.3 glass is an essential tool for efficient filtration and separation processes in laboratory applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)