Key Documents
BR458021
BRAND® filter crucible
borosilicate glass, capacity 30 mL, 1 D 1
Synonym(s):
Filter, Filter crucible
Select a Size
10 EA
$417.00
$417.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
10 EA
$417.00
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
$417.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
manufacturer/tradename
BRAND 458021
capacity
30 mL
pore size
100-160 μm
General description
The BRAND® Filter Crucible made from Boro 3.3 glass is an essential tool for efficient filtration and separation processes in laboratory applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)