BR458023
BRAND® filter crucible
borosilicate glass, capacity 30 mL, 1 D 3
Synonym(s):
Filter, Filter crucible
Select a Size
$417.00
Select a Size
About This Item
$417.00
Recommended Products
material
borosilicate glass
manufacturer/tradename
BRAND 458023
capacity
30 mL
pore size
16-40 μm
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)