BR23882
BRAND® support for compact automatic burette
polypropylene base, size 210 mm × 55 mm
Synonym(s):
Accessories, Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
Select a Size
$176.00
Select a Size
About This Item
$176.00
Recommended Products
material
polypropylene base
stainless steel rod
packaging
pack of 1 ea
manufacturer/tradename
BRAND 23882
base size
210 mm × 155 mm
rod size
505 mm × 12 mm
size
210 mm × 55 mm
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service