Key Documents
BR578000
BRAND® burette clamp, aluminum die casting for 2 burettes, with roll mounting
Synonym(s):
Accessories, Burette clamp, Burettes
Select a Size
1 EA
$122.00
$122.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$122.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.43
$122.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
BRAND 78000
Related Categories
General description
Burettes up to 50 ml can be mounted in seconds. PVC-coated clamps provide a secure grip. Comfortable height adjustment. Graduations remain visible. Very sturdy design, easy to operate. Integral support connector.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)