Key Documents
Z691453
Scienceware® compact support stand
center and side mounts, base W × L 8 1/4 in. × 6 1/4 in., rod diam. × L 3/8 in. × 19 3/4 in.
Select a Size
1 EA
$105.00
$105.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$105.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
$105.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene base
stainless steel rod
manufacturer/tradename
Bel-Art H18303-0005
base W × L
8 1/4 in. × 6 1/4 in.
rod diam. × L
3/8 in. × 19 3/4 in.
General description
This non-corrosive, molded support stand with stainless steel rod saves bench space. Lighter and less expensive than metal ring stands, it can be used to support a variety of types of plasticware and labware. The compact base has a smooth surface for easy cleaning and weighs 1.95 kg (4.3 lb) providing excellent stability. The stainless steel rod can be mounted at the center or at the end of the base. The base is made of corrosion-resistant polypropylene and is autoclavable.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1