Skip to Content
Merck
All Photos(1)

Key Documents

912999

Sigma-Aldrich

Silica dioxide- vs nanofiber

Synonym(s):

Nanofibrous inorganic powder structure

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$150.00
10 G
$223.00

$150.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$150.00
10 G
$223.00

About This Item

Linear Formula:
SiO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

$150.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

form

nanofiber

Quality Level

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

Inorganic nanofibers have unique properties (i.e. high surface area, high porosity, good breathability, large surface to volume ratio, and stability) their applications include anodes and cathodes in li-ion batteries. The can additionally act as a fuel cell separator, catalyst, catalyst support, photo catalyst, gas sensors, thermal insulators, metal or ceramic nano-composites, dehumidifiers, abrasives, thermal barrier coatings, and filtration.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What does the "vs" in the name of product 912999 refer to?

    1 answer
    1. The "vs" in SiO2 VS stands for SiO2 that utilizes water glass Na2SiO3. This type of SiO2 does not have nearly as many surfaces as pure SiO2 and may contain up to 1% Na.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service