Skip to Content
Merck
All Photos(1)

Key Documents

914150

Sigma-Aldrich

Silica dioxide- sorbent milled nanofiber

Synonym(s):

Nanofibrous inorganic powder structure, SiO2

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$338.00
10 G
$588.00

$338.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$338.00
10 G
$588.00

About This Item

Empirical Formula (Hill Notation):
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
NACRES:
NA.23

$338.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

form

nanofiber

Quality Level

fiber size

300-800 nm , structure: amorphous

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

Silica dioxide- sorbent nanofibers can absorb significantly more water than commercially aailable silicagel of the same mesoporous character. Applications include adsorbent of water and other polar sorbents, catalyst carrier, filtration, separation, li-ion battery separators, and sensors.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service