Skip to Content
Merck
All Photos(1)

Key Documents

913499

Sigma-Aldrich

Aluminum oxide nanofibers

Synonym(s):

Al2O3, Nanofibrous inorganic powder

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$222.00
10 G
$694.00

$222.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$222.00
10 G
$694.00

About This Item

Linear Formula:
Al2O3
CAS Number:
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.23

$222.00


Only 0 left in stock

Request a Bulk Order

description

Thermal Conductivity 30 Wm-1K-1

Quality Level

form

nanofiber
powder (3D cotton/white fluffy)

fiber diam. × L

300-900 nm ± 100 × 2-hundreds of μm , Structure: Polycystalline nanofibers

surface area

40-70 m2/g

primary crystallite size

(gamma

Typical Size of Crystallites:N/A)

conductivity

<10^-16 S/cm (Electrical )

mp

2072 °C

SMILES string

[Al+3].[Al+3].[O-2].[O-2].[O-2]

InChI

1S/2Al.3O/q2*+3;3*-2

InChI key

PNEYBMLMFCGWSK-UHFFFAOYSA-N

Application

Gamma Alumina has applications in optical displays, catalyst and catalyst carrier, electrical components- electrical insulator, thermal conductor, nanocomposites, separator, separator material for batteries.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service