Skip to Content
Merck
All Photos(1)

Key Documents

922587

Sigma-Aldrich

Silicon Dioxide (Silica) - Nanofibers

diam. × L ~425 nm (±50 nm) × infinity

Synonym(s):

Nanofibrous inorganic powder, SiO2

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$220.00

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$220.00

About This Item

Empirical Formula (Hill Notation):
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

diam. × L

~425 nm (±50 nm) × infinity

Quality Level

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

Silica nanofibers are yielded through a novel ac electrospinning process resulting in extraordinary aspect ratios, consistent purity, and consistent certification of form parameters that are difficult if not impossible to achieve using other nanofiber production methods. Potential applications include filtration membranes and catalyst support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service