Skip to Content
Merck
All Photos(1)

Key Documents

764205

Sigma-Aldrich

Alkyne-PEG4-maleimide

90% (HPLC)

Synonym(s):

Acetylene-PEG4-maleimide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$99.50
25 MG
$210.00

$99.50


Estimated to ship onApril 12, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$99.50
25 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C18H26N2O7
Molecular Weight:
382.41
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

$99.50


Estimated to ship onApril 12, 2025


Request a Bulk Order

Quality Level

Assay

90% (HPLC)

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

mp

62-67 °C

functional group

alkyne
maleimide

storage temp.

−20°C

SMILES string

O=C(N1CCC(NCCOCCOCCOCCOCC#C)=O)C=CC1=O

InChI

1S/C18H26N2O7/c1-2-8-24-10-12-26-14-15-27-13-11-25-9-6-19-16(21)5-7-20-17(22)3-4-18(20)23/h1,3-4H,5-15H2,(H,19,21)

InChI key

GHEBECZRHJXPTL-UHFFFAOYSA-N

Application

Alkyne-PEG4-maleimide has been used for modification of Csy containing bacterial collagens by introducing alkyne moiety to enable it for further chemical modification.[1]
Heterobifunctional crosslinker enabling the connection of an azide containing compound or biomolecule to one with a thiol. The maleimide group will react preferentially with thiols to form a covalent adduct and the terminal alkyne will react with azides via copper-catalyzed 1,3-dipolar cycloaddition click chemistry. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification cysteine containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering specific chemical modification sites into a collagen-like protein from Streptococcus pyogenes.
Stoichevska V, et al.
Journal of Biomedical Materials Research Part A (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service