Skip to Content
Merck
All Photos(2)

Key Documents

900941

Sigma-Aldrich

Chloro(4-cyanophenyl)[(R)-1-[(S)-2-[bis(4-fluorophenyl]phosphino]ferrocenyl]ethyldi-tert-butylphosphine]nickel(II)

≥95%

Synonym(s):

SK-J014-1n

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$299.00

$299.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$299.00

About This Item

Empirical Formula (Hill Notation):
C39H42ClF2FeNNiP2
CAS Number:
Molecular Weight:
774.69
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$299.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or solid

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

SMILES string

CC(C)(C)P(C(C)(C)C)C[C]1[C][C][C][C]1P(C2=CC=C(F)C=C2)C3=CC=C(F)C=C3.Cl[Ni]C4=CC=C(C#N)C=C4.[C]5[C][C][C][C]5.[Fe]

Application

Amination of functionalized aryl chlorides with ammonia and ammonium salts.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluating 1, 1?-Bis (phosphino) ferrocene Ancillary Ligand Variants in the Nickel-Catalyzed C?N Cross-Coupling of (Hetero) aryl Chlorides.
Clark J S, et al.
Organometallics, 36(3), 679-686 (2017)
Nickel?Catalyzed Monoarylation of Ammonia.
Borzenko A, et al.
Angewandte Chemie (International Edition in English), 54(12), 3773-3777 (2015)
Ni?Catalyzed Amination Reactions: An Overview.
Marin M, et al.
Chemical Record, 16(4), 1819-1832 (2016)
Nickel?Catalyzed Amination of Aryl Chlorides with Ammonia or Ammonium Salts.
Green R A and Hartwig J F
Angewandte Chemie (International Edition in English), 54(12), 3768-3772 (2015)
Sarah Z Tasker et al.
Nature, 509(7500), 299-309 (2014-05-16)
Tremendous advances have been made in nickel catalysis over the past decade. Several key properties of nickel, such as facile oxidative addition and ready access to multiple oxidation states, have allowed the development of a broad range of innovative reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service