Skip to Content
Merck
All Photos(2)

Documents

900943

Sigma-Aldrich

Chloro(4-cyanophenyl)[(R)-1-[(S)-2-(dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine]nickel(II)

Synonym(s):

SK-J003-1n

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H60ClFeNNiP2
CAS Number:
Molecular Weight:
802.88
UNSPSC Code:
12352103
NACRES:
NA.22

form

powder or solid

Quality Level

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst
reaction type: Asymmetric synthesis

SMILES string

Cl[Ni+2]C1=CC=C(C#N)C=C1.C[C@@H](P(C2CCCCC2)C3CCCCC3)[C]4[C][C][C][C]4P(C5CCCCC5)C6CCCCC6.[C]7[C][C][C][C]7.[Fe]

Application

Amination of aryl chlorides, bromides and sulfonates with ammonia. Amination of aryl carbamates with ammonia or ammonium salts.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluating 1, 1?-Bis (phosphino) ferrocene Ancillary Ligand Variants in the Nickel-Catalyzed C?N Cross-Coupling of (Hetero) aryl Chlorides.
Clark J S, et al.
Organometallics, 36(3), 679-686 (2017)
Nickel?Catalyzed Monoarylation of Ammonia.
Borzenko A, et al.
Angewandte Chemie (International Edition in English), 54(12), 3773-3777 (2015)
Nickel?Catalyzed Amination of Aryl Chlorides with Ammonia or Ammonium Salts.
Green R A and Hartwig J F
Angewandte Chemie (International Edition in English), 54(12), 3768-3772 (2015)
Ni?Catalyzed Amination Reactions: An Overview.
Marin M, et al.
Chemical Record, 16(4), 1819-1832 (2016)
Sarah Z Tasker et al.
Nature, 509(7500), 299-309 (2014-05-16)
Tremendous advances have been made in nickel catalysis over the past decade. Several key properties of nickel, such as facile oxidative addition and ready access to multiple oxidation states, have allowed the development of a broad range of innovative reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service