Skip to Content
Merck
All Photos(1)

Key Documents

900853

Sigma-Aldrich

EH-IDTBR

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$459.00

$459.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$459.00

About This Item

Empirical Formula (Hill Notation):
C72H88N6O2S8
CAS Number:
Molecular Weight:
1326.03
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

$459.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Band gap: 1.68 eV

Quality Level

Assay

97%

form

solid

solubility

chloroform: soluble
dichlorobenzene: soluble

Orbital energy

HOMO -5.58 eV 
LUMO -3.9 eV 

SMILES string

S1\C(=C/c2c3n[s]nc3c(cc2)c4[s]c5c(c4)C(c6c5cc7c(c6)c8[s]c(cc8C7(CC(CCCC)CC)CC(CCCC)CC)c9c%10n[s]nc%10c(cc9)\C=C%11/SC(=S)N(C/%11=O)CC)(CC(CCCC)CC)CC(CCCC)CC)\C(=O)N(C1=S)CC

InChI

1S/C72H88N6O2S8/c1-11-21-25-43(15-5)39-71(40-44(16-6)26-22-12-2)53-35-52-54(36-51(53)65-55(71)37-57(83-65)49-31-29-47(61-63(49)75-87-73-61)33-59-67(79)77(19-9)69(81)85-59)72(41-45(17-7)27-23-13-3,42-46(18-8)28-24-14-4)56-38-58(84-66(52)56)50-32-30-48(62-6

InChI key

HKJHYTKBDDVGRK-IOUDQCQMSA-N

Application

EH-IDTBR is a branched non-fullerene acceptor (NFAs) used in organic photovoltaic (OPV) devices. It is used along with donor polymer, poly (3-hexylthiophene) (P3HT) to fabricate the solar cells to enhance the power conversion efficiencies and stabilities of the devices.[1][2] It is also used to develop n-type small molecules for organic thin-film transistors (OTFTs) because of their n-type electron mobility.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterisation of non-fullerene electron acceptors for organic photovoltaics
Thesis (2018)
Impact of Nonfullerene Acceptor Side Chain Variation on Transistor Mobility
Bristow H, et al.
Advanced Electronic Materials, 5(10), 1900344-1900344 (2019)
Sarah Holliday et al.
Nature communications, 7, 11585-11585 (2016-06-10)
Solution-processed organic photovoltaics (OPV) offer the attractive prospect of low-cost, light-weight and environmentally benign solar energy production. The highest efficiency OPV at present use low-bandgap donor polymers, many of which suffer from problems with stability and synthetic scalability. They also
Charge recombination losses in non-fullerene acceptor based organic solar cells
Thesis (2018)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service