Skip to Content
Merck
All Photos(1)

Key Documents

900810

Sigma-Aldrich

O-IDTBR

Synonym(s):

IDTBR-O

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$542.00
100 MG
$863.00

$542.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$542.00
100 MG
$863.00

About This Item

Empirical Formula (Hill Notation):
C72H88N6O2S8
Molecular Weight:
1326.03
UNSPSC Code:
12352200
NACRES:
NA.23

$542.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Band gap: 1.63 eV

Quality Level

Assay

99%

form

solid

solubility

organic solvents: soluble

Orbital energy

HOMO -5.51 eV 
LUMO -3.88 eV 

SMILES string

S=C(N(CC)C/1=O)SC1=C/C(C2=NSN=C23)=CC=C3C4=CC5=C(S4)C6=CC7=C(C(SC(C8=CC=C(/C=C9SC(N(CC)C\9=O)=S)C%10=NSN=C%108)=C%11)=C%11C7(CCCCCCCCC)CCCCCCCCC)C=C6C5(CCCCCCCCC)CCCCCCCCC

InChI

1S/C76H96N6O2S8/c1-7-13-17-21-25-29-33-41-75(42-34-30-26-22-18-14-8-2)57-47-56-58(48-55(57)69-59(75)49-61(87-69)53-39-37-51(65-67(53)79-91-77-65)45-63-71(83)81(11-5)73(85)89-63)76(43-35-31-27-23-19-15-9-3,44-36-32-28-24-20-16-10-4)60-50-62(88-70(56)60)54-

InChI key

VUPVAPDLDBEWMY-GFPJCWIKSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient and thermally stable fullerene-free organic solar cells based on a small molecule donor and acceptor.
Sachin B, et al.
Journal of Material Chemistry A, 4(42), 16335-16340 (2016)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service